N-Succinimidyl Acrylate 97%

Supplier: Ambeed
Warning

Synonyms: N-Acryloxysuccinimide, 2,5-Dioxopyrrolidin-1-yl acrylate, N-Acryloyloxysuccinimide, Acrylic Acid N-Succinimidyl Ester

A107972-10G A107972-1G A107972-250MG A107972-5G A107972-25G
77188-906EA 56.65 USD
77188-906 77188-908 77188-910 77188-912 77549-088
N-Succinimidyl Acrylate 97%
N-Succinimidyl Acrylate

Caution: for Research Use Only

Formula: C₇H₇NO₄
MW: 169.14 g/mol
MDL Number: MFCD00078261
CAS Number: 38862-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR