Phenylphosphonic acid 98%

Supplier: Ambeed
Danger

Synonyms: Phenylphosphonic Acid

A944580-100G A944580-25G A944580-5G
77159-970EA 45 USD
77159-970 77159-972 77159-974
Phenylphosphonic acid 98%
Phenylphosphonic acid

Caution: for Research Use Only

Formula: C₆H₇O₃P
MDL Number: MFCD00002136
CAS Number: 1571-33-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR