Sodium tetraphenylborate 98%

Supplier: Ambeed
Danger

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

A696394-10G A696394-25G A696394-5G A696394-100G
77158-406EA 8 USD
77158-406 77158-408 77158-410 77901-529
Sodium tetraphenylborate 98%
Sodium tetraphenylborate

Caution: for Research Use Only

Formula: (C₆H₅)₄BNa
MDL Number: MFCD00011494
CAS Number: 143-66-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR