Cobalt(II) acetylacetonate 96%

Supplier: Ambeed
Danger

Synonyms: Cobalt(II) bis(acetylacetonate), Cobalt (II) bis(2,4-pentanedionato-O,O'), Cobalt (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')cobalt (II)

A694568-25G A694568-500G A694568-100G
77158-404EA 14 USD
77158-404 77614-148 77686-678
Cobalt(II) acetylacetonate 96%
Cobalt (II) acetylacetonate

Caution: for Research Use Only

Formula: C₁₀H₁₄CoO₄
MDL Number: MFCD00000014
CAS Number: 14024-48-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR