Fast green FCF 95%

Supplier: Ambeed
Danger

A670295-25MG A670295-1G A670295-5G
77158-148EA 5 USD
77158-148 77900-886 77901-049
Fast green FCF 95%
Fast Green FCF

Caution: for Research Use Only

Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
MDL Number: MFCD00013053
CAS Number: 2353-45-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR