1-Ethynylpyrene 98%

Supplier: Ambeed
Warning

A496061-250MG A496061-1G A496061-5G
77156-620EA 16 USD
77156-620 77686-299 77900-745
1-Ethynylpyrene 98%
1-Ethynylpyrene

Caution: for Research Use Only

Formula: C₁₈H₁₀
MW: 226.28 g/mol
MDL Number: MFCD02093933
CAS Number: 34993-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR