4-Bromotetrafluoroaniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,3,5,6-tetrafluoroaniline

A487375-1G A487375-250MG A487375-5G
77156-522EA 21 USD
77156-522 77156-524 77156-526
4-Bromotetrafluoroaniline 98%
4-Bromotetrafluoroaniline

Caution: for Research Use Only

Formula: C₆H₂BrF₄N
MW: 243.9844 g/mol
MDL Number: MFCD00012144
CAS Number: 1998-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR