Pentacene 99%, purified by sublimation

Supplier: Ambeed
Warning

Synonyms: 2,3,6,7-Dibenzanthracene

A426110-100MG A426110-1G A426110-250MG A426110-5G
77155-860EA 32.96 USD
77155-860 77155-862 77155-864 77686-168
Pentacene 99%, purified by sublimation
Pentacene

Caution: for Research Use Only

Formula: C₂₂H₁₄
MW: 278.35 g/mol
MDL Number: MFCD00003710
CAS Number: 135-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR