(Ferrocenylmethyl)trimethylammonium iodide 97%

Supplier: Ambeed
Warning

Synonyms: (Ferrocenylmethyl)trimethylammonium Iodide, Ferrocenylmethyltrimethylammonium iodide

A384261-250MG A384261-5G
77155-372EA 14 USD
77155-372 77155-374
(Ferrocenylmethyl)trimethylammonium iodide 97%
(Ferrocenylmethyl)trimethylammonium iodide

Caution: for Research Use Only

Formula: C₁₄H₂₀FeIN
MDL Number: MFCD00041534
CAS Number: 12086-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR