Filgotinib (GLPG0634) 99%

Supplier: Ambeed
Warning

Synonyms: N-(5-(4-((1,1-Dioxidothiomorpholino)methyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide, Filgotinib

A302163-10MG A302163-1MG A302163-5MG A302163-100MG A302163-1G A302163-250MG
77154-198EA 22 USD
77154-198 77154-200 77154-202 77685-862 77685-863 77685-864
Filgotinib (GLPG0634) 99%
Filgotinib (GLPG0634)

Caution: for Research Use Only

Formula: C₂₁H₂₃N₅O₃S
MDL Number: MFCD20527867
CAS Number: 1206161-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR