Inosine 5'-monophosphate disodium salt hydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate
A294770-25G
A294770-5G
A294770-100G
A294770-500G
77154-102EA
20
USD
77154-102
77154-104
77612-694
77685-838
Inosine 5'-monophosphate disodium salt hydrate 97%
Inosine 5'-monophosphate disodium salt hydrate
Caution: for Research Use Only
Formula:
C₁₀H₁₁N₄Na₂O₈P·xH₂O MW: 392.17 g/mol (anhy) |
MDL Number:
MFCD00150372 CAS Number: 352195-40-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...