LEE011 98%

Supplier: Ambeed
Warning

Synonyms: Ribociclib, 7-Cyclopentyl-N,N-dimethyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)-7H-pyrrolo[2,3-d]pyrimidine-6-carboxamide

A288696-10MG A288696-1G A288696-1MG A288696-250MG A288696-5MG A288696-25G A288696-50MG A288696-5G
77153-954EA 17 USD
77153-954 77550-766 77612-662 77612-664 77612-666 77685-823 77901-722 77900-799
LEE011 98%
LEE011

Caution: for Research Use Only

Formula: C₂₃H₃₀N₈O
MDL Number: MFCD27976795
CAS Number: 1211441-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR