RuCl[(R,R)-Tsdpen](p-cymene) 98%

Supplier: Ambeed
Warning

Synonyms: Chloro(p-cymene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II), [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), RuCl(p-cymene)[(R,R)-Ts-DPEN], Chloro(p-cymene)[(R,R)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), Chloro{[(1R,2R)-(-)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II), Chloro{[(1R,2R)-(-)-2-amino-1,2-diphenylethyl](4-toluenesulphonyl)amido}(p-cymene)ruthenium(II), [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II)

A233712-1G A233712-250MG A233712-5G A233712-100G A233712-25G
77152-946EA 5.15 USD
77152-946 77152-948 77152-950 77152-952 77612-284 77612-286
RuCl[(R,R)-Tsdpen](p-cymene) 98%
RuCl[(R,R)-Tsdpen](p-cymene)

Caution: for Research Use Only

Formula: C₃₁H₃₅ClN₂O₂RuS
MDL Number: MFCD10567385
CAS Number: 192139-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR