Veratraldehyde 98%

Supplier: Ambeed
Warning

A186120-100G A186120-25G A186120-500G A186120-1000G
77151-786EA 19 USD
77151-786 77151-788 77151-790 77611-798
Veratraldehyde 98%
Veratraldehyde

Caution: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00003363
CAS Number: 120-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR