JSH-23 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methyl-N1-(3-phenylpropyl)-1,2-phenylenediamine, JSH-23, 4-Methyl-N1-(3-phenylpropyl)benzene-1,2-diamine
A153850-10MG
A153850-1MG
A153850-5MG
A153850-100MG
A153850-250MG
77150-508EA
28.84
USD
77150-508
77150-510
77150-512
77685-273
77685-274
JSH-23 98%
JSH-23
Caution: for Research Use Only
Formula:
C₁₆H₂₀N₂ MW: 240.3 g/mol |
MDL Number:
MFCD09753595 CAS Number: 749886-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...