MLN0128 98%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)benzo[d]oxazol-2-amine, INK-128, Sapanisertib

A121801-10MG A121801-2MG A121801-5MG A121801-1MG A121801-100MG A121801-50MG A121801-250MG
77149-222EA 77 USD
77149-222 77149-224 77149-226 77611-036 77685-074 77685-075 77901-630
MLN0128 98%
MLN0128

Caution: for Research Use Only

Formula: C₁₅H₁₅N₇O
MDL Number: MFCD22124893
CAS Number: 1224844-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR