AV-412 98+%

Supplier: Ambeed
Warning

A116496-10MG A116496-1MG A116496-25MG A116496-2MG A116496-5MG A116496-50MG
77148-960EA 41 USD
77148-960 77148-962 77148-964 77148-966 77148-968 77685-031
AV-412 98+%
AV-412

Caution: for Research Use Only

Formula: C₂₇H₂₈ClFN₆O
MDL Number: MFCD16038939
CAS Number: 451492-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR