Ethyl-2-phenylacrylate 97% stabilised

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-phenylacrylate

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A570354-250MG A570354-1G
77144-636EA 83.43 USD
77144-636 77686-455
Ethyl-2-phenylacrylate 97% stabilised
Ethyl-2-phenylacrylate

Caution: for Research Use Only

Formula: C₁₁H₁₂O₂
MDL Number: MFCD00026862
CAS Number: 22286-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR