(-)-Perillyl alcohol 95% stabilized

Supplier: Ambeed
Warning

Synonyms: (S)-4-Isopropenyl-1-cyclohexenylmethanol, (S)-p-Mentha-1,8-dien-7-ol, (S)-(4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl)methanol, (S)-(−)-Perillyl alcohol

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A370070-1G A370070-250MG A370070-5G
77142-440EA 19.1 USD
77142-440 77142-442 77142-444
(-)-Perillyl alcohol 95% stabilized
(-)-Perillyl alcohol

Caution: for Research Use Only

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
MDL Number: MFCD00062995
CAS Number: 18457-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR