(R,R)-(-)-3,6-Octanediol 98%

Supplier: Ambeed
Warning

Synonyms: (3R,6R)-Octane-3,6-diol, (R,R)-(-)-3,6-Dihydroxy octane, (3R,6R)-(-)-3,6-Octanediol

A210300-100MG A210300-1G A210300-250MG
77139-578EA 18.54 USD
77139-578 77139-580 77139-582
(R,R)-(-)-3,6-Octanediol 98%
(R,R)-(-)-3,6-Octanediol

Caution: for Research Use Only

Formula: C₈H₁₈O₂
MDL Number: MFCD01631087
CAS Number: 129619-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR