5-Amino-2-chlorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A159259-100G
A159259-10G
A159259-25G
A159259-5G
77138-062EA
133.68
USD
77138-062
77138-064
77138-066
77138-068
5-Amino-2-chlorobenzoic acid 98%
5-Amino-2-chlorobenzoic acid
Caution: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol |
MDL Number:
MFCD00007845 CAS Number: 89-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...