GDC-0032 98+%

Supplier: Ambeed
Warning

Synonyms: Taselisib, 2-(4-(2-(1-Isopropyl-3-methyl-1H-1,2,4-triazol-5-yl)-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepin-9-yl)-1H-pyrazol-1-yl)-2-methylpropanamide

A139829-10MG A139829-1MG A139829-50MG A139829-5MG
77137-258EA 47.39 USD
77137-258 77137-260 77137-262 77137-264
GDC-0032 98+%
GDC-0032

Caution: for Research Use Only

Formula: C₂₄H₂₈N₈O₂
MDL Number: MFCD28167893
CAS Number: 1282512-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR