5-Chlorosalicylaldehyde 97%

Supplier: Ambeed
Warning

A129742-100G A129742-10G A129742-25G A129742-500G A129742-5G
77136-866EA 41.2 USD
77136-866 77136-868 77136-870 77136-872 77136-874
5-Chlorosalicylaldehyde 97%
5-Chlorosalicylaldehyde

Caution: for Research Use Only

Formula: C₇H₅ClO₂
MDL Number: MFCD00003331
CAS Number: 635-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR