Ethyl 2-amino-5-fluorobenzoate 97%
Supplier: Ambeed
|
Warning
|
A123283-100MG
A123283-10G
A123283-1G
A123283-250MG
77136-592EA
5.15
USD
77136-592
77136-594
77136-596
77136-598
Ethyl 2-amino-5-fluorobenzoate 97%
Ethyl 2-amino-5-fluorobenzoate
Caution: for Research Use Only
Formula:
C₉H₁₀FNO₂ |
MDL Number:
MFCD11042774 CAS Number: 391-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...