Ethylene carbonate 99%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dioxa-2-cyclopentanone

A109810-100G A109810-500G A109810-1000G
77135-938EA 6.37 USD
77135-938 77135-940 77549-120
Ethylene carbonate 99%
Ethylene carbonate

Caution: for Research Use Only

Formula: C₃H₄O₃
MW: 88.06 g/mol
MDL Number: MFCD00005382
CAS Number: 96-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR