SMI-4a 98%

Supplier: Ambeed
Warning

A100114-10MG A100114-1MG A100114-50MG A100114-5MG
77135-406EA 75.19 USD
77135-406 77135-408 77135-410 77135-412
SMI-4a 98%
SMI-4a

Caution: for Research Use Only

Formula: C₁₁H₆F₃NO₂S
MDL Number: MFCD01152003
CAS Number: 327033-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR