Theophyllin 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline

A896968-1MG A896968-1G A896968-5G
77118-762EA 5.31 USD
77118-762 77614-608 77686-938
Theophyllin 98%
Theophyllin

Caution: for Research Use Only

Formula: C₇H₈N₄O₂
MW: 180.16 g/mol
MDL Number: MFCD00079619
CAS Number: 58-55-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR