1-Bromo-2-fluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromofluorobenzene

A868004-10G A868004-25G
77118-638EA 5.15 USD
77118-638 77118-640
1-Bromo-2-fluorobenzene 98%
1-Bromo-2-fluorobenzene

Caution: for Research Use Only

Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155
MDL Number: MFCD00000282
CAS Number: 1072-85-1
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR