Sodium palmitate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Palmitic Acid Sodium Salt
A607579-100G
A607579-1G
A607579-25G
A607579-500G
A607579-5G
A607579-1000G
77117-328EA
46.68
USD
77117-328
77117-330
77117-332
77117-334
77117-336
77686-536
Sodium palmitate 98%
Sodium palmitate
Caution: for Research Use Only
Formula:
C₁₆H₃₂O₂ MW: 278.41 g/mol |
MDL Number:
MFCD00002749 CAS Number: 408-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...