Pyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

A544876-100G A544876-25G A544876-500G A544876-1000G
77116-838EA 20.6 USD
77116-838 77116-840 77116-842 77551-650
Pyridine-2-carbonitrile 98%
Pyridine-2-carbonitrile

Caution: for Research Use Only

Formula: C₆H₄N₂
MDL Number: MFCD00006218
CAS Number: 100-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR