1,3,2-Dioxathiolane-2-oxide 98%
Supplier: Ambeed
|
Danger
|
A461480-25G
A461480-5G
77116-290EA
8.49
USD
77116-290
77116-292
77116-294
1,3,2-Dioxathiolane-2-oxide 98%
1,3,2-Dioxathiolane-2-oxide
Caution: for Research Use Only
Formula:
C₂H₄O₃S |
MDL Number:
MFCD00005354 CAS Number: 3741-38-6 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...