4-Methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Hydroxymethyl)toluole, p-Tolylmethanol, (4-Methylphenyl)methanol

A261012-100G A261012-25G A261012-5G
77113-680EA 45.32 USD
77113-680 77113-682 77113-684
4-Methylbenzyl alcohol 98%
4-Methylbenzyl alcohol

Caution: for Research Use Only

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 217
Melting Pt: 59
MDL Number: MFCD00004664
CAS Number: 589-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR