Psoralen 98+%

Supplier: Ambeed
Warning

A232760-100MG A232760-10MG A232760-1G A232760-1MG A232760-250MG A232760-50MG A232760-5MG A232760-5G
77113-042EA 54.59 USD
77113-042 77113-044 77113-046 77113-048 77113-050 77113-052 77113-054 77685-643
Psoralen 98+%
Psoralen

Caution: for Research Use Only

Formula: C₁₁H₆O₃
MDL Number: MFCD00010520
CAS Number: 66-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR