5-Acetyluracil 98%

Supplier: Ambeed
Warning

Synonyms: 5-Acetylpyrimidine-2,4(1H,3H)-dione

A227342-10G A227342-1G A227342-25G A227342-5G
77112-880EA 31.94 USD
77112-880 77112-882 77112-884 77112-886
5-Acetyluracil 98%
5-Acetyluracil

Caution: for Research Use Only

Formula: C₆H₆N₂O₃
MDL Number: MFCD00151968
CAS Number: 6214-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR