Adefovir dipivoxil 98%

Supplier: Ambeed
Warning

Synonyms: Adefovir Dipivoxil, 9-(2[bis(Pivaloyloxymethoxy)phosphorylmethoxy]ethyl)adenine, 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine, ((((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphoryl)bis(oxy))bis(methylene) bis(2,2-dimethylpropanoate)

A209543-10MG A209543-5G A209543-1G A209543-1MG
77112-414EA 6.18 USD
77112-414 77615-762 77615-760 77685-543
Adefovir dipivoxil 98%
Adefovir dipivoxil

Caution: for Research Use Only

Formula: C₂₀H₃₂N₅O₈P
MW: 501.48 g/mol
MDL Number: MFCD00869897
CAS Number: 142340-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR