3,4-Dimethoxybenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol

A187224-100G A187224-10G A187224-1G A187224-25G A187224-500G A187224-5G
77111-610EA 57.68 USD
77111-610 77111-612 77111-614 77111-616 77111-618 77111-620
3,4-Dimethoxybenzyl alcohol 97%
3,4-Dimethoxybenzyl alcohol

Caution: for Research Use Only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00004638
CAS Number: 93-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR