Methylurea 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylurea, N-Methylurea, Methylcarbamide

A173396-1000G A173396-100G A173396-25G A173396-500G
77111-104EA 32.96 USD
77111-104 77111-106 77111-108 77111-110
Methylurea 97%
Methylurea

Caution: for Research Use Only

Formula: C₂H₆N₂O
MW: 74.08 g/mol
Boiling Pt: 240
MDL Number: MFCD00007950
CAS Number: 598-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR