Glycoluril 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione, Acetylenediurea
A153729-1000G
A153729-100G
A153729-10G
A153729-25G
A153729-500G
77110-418EA
75.18
USD
77110-418
77110-420
77110-422
77110-424
77110-426
Glycoluril 97%
Glycoluril
Caution: for Research Use Only
Formula:
C₄H₆N₄O₂ MW: 142.12 g/mol |
MDL Number:
MFCD00005539 CAS Number: 496-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...