2,3,4,5-Tetrafluorobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A143918-1000G
A143918-100G
A143918-10G
A143918-25G
A143918-500G
A143918-5G
77109-912EA
145.23
USD
77109-912
77109-914
77109-916
77109-918
77109-920
77109-922
2,3,4,5-Tetrafluorobenzoic acid 95%
2,3,4,5-Tetrafluorobenzoic acid
Caution: for Research Use Only
Formula:
C₇H₂F₄O₂ |
MDL Number:
MFCD00009613 CAS Number: 1201-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...