Guaifenesin 98%

Supplier: Ambeed
Warning

A113558-100G A113558-10G A113558-1MG A113558-25G
77108-132EA 30.91 USD
77108-132 77108-134 77108-136 77108-138
Guaifenesin 98%
Guaifenesin

Caution: for Research Use Only

Formula: C₁₀H₁₄O₄
MDL Number: MFCD00016873
CAS Number: 93-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR