3-Fluoropyridine 98%

Supplier: Ambeed
Danger

A290551-25G A290551-5G
77099-302EA 5.31 USD
77099-302 77099-304 77099-306
3-Fluoropyridine 98%
3-Fluoropyridine

Caution: for Research Use Only

Formula: C₅H₄FN
MDL Number: MFCD00006374
CAS Number: 372-47-4
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR