Sodium salicylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt
A198464-1000G
A198464-100G
A198464-500G
A198464-50MG
77097-354EA
20.16
USD
77097-354
77097-356
77097-358
77685-494
Sodium salicylate 95%
Sodium salicylate
Caution: for Research Use Only
Formula:
HOC₆H₄COONa MW: 160.10 g/mol |
MDL Number:
MFCD00002440 CAS Number: 54-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...