DL-Alanine 98,5%

Supplier: Ambeed
Warning

A129900-100G A129900-25G
77094-926EA 13 USD
77094-926 77094-928
DL-Alanine 98,5%
DL-Alanine

Caution: for Research Use Only

Formula: C₃H₇NO₂
MDL Number: MFCD00064408
CAS Number: 302-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR