Trimethylolpropane triacrylate 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 200 ppm

A111327-100G A111327-25G A111327-500G
77094-012EA 19 USD
77094-012 77094-014 77094-016
Trimethylolpropane triacrylate 95% stabilized
Trimethylolpropane triacrylate

Caution: for Research Use Only

Formula: C₁₅H₂₀O₆
MDL Number: MFCD00008628
CAS Number: 15625-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR