3-Amino-5-methylcaproic acid 98%

Supplier: Ambeed
Warning

Synonyms: (+/-)-3-Amino-5-methylhexanoic acid, DL-beta-Homoleucine, 3-Amino-5-methylhexanoic acid

A107165-1G A107165-250MG A107165-5G
77093-790EA 21.22 USD
77093-790 77093-792 77093-794
3-Amino-5-methylcaproic acid 98%
3-Amino-5-methylcaproic acid

Caution: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
MDL Number: MFCD01863281
CAS Number: 3653-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR