9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%

Supplier: Ambeed
Danger

A945183-10G A945183-25G A945183-5G
77007-158EA 11 USD
77007-158 77007-160 77007-162
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%
9-Fluorenylmethyl chloroformate (Fmoc chloride)

Caution: for Research Use Only

Formula: C₁₅H₁₁ClO₂
MDL Number: MFCD00001138
CAS Number: 28920-43-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR