(4-(1-Aminocyclopropyl)phenyl)boronic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A811619-100MG
A811619-250MG
A811619-1G
A811619-50MG
A811619-5G
77019-464EA
37
USD
77019-464
77019-466
77891-559
77891-560
77891-561
(4-(1-Aminocyclopropyl)phenyl)boronic acid hydrochloride 97%
(4-(1-Aminocyclopropyl)phenyl)boronic acid hydrochloride
Caution: for Research Use Only
Formula:
C₉H₁₃BClNO₂ |
MDL Number:
MFCD16660279 CAS Number: 1256345-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...