6-O-Stearoyl-L-ascorbic acid 98%

Supplier: Ambeed
Warning

Synonyms: L-Ascorbyl 6-Stearate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl stearate

A708872-100MG A708872-1G A708872-250MG A708872-5G
77006-036EA 13.8 USD
77006-036 77006-038 77006-040 77006-042
6-O-Stearoyl-L-ascorbic acid 98%
6-O-Stearoyl-L-ascorbic acid

Caution: for Research Use Only

Formula: C₂₄H₄₂O₇
MW: 442.59 g/mol
MDL Number: MFCD00059739
CAS Number: 10605-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR