3,4,5-Trifluoroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1,2,3-Trifluoro-5-methoxybenzene, Trifluoro-3,4,5-anisole, 1-Methoxy-3,4,5-trifluorobenzene

A691792-1G A691792-25G A691792-5G
77005-930EA 21 USD
77005-930 77005-932 77005-934
3,4,5-Trifluoroanisole 97%
3,4,5-Trifluoroanisole

Caution: for Research Use Only

Formula: C₇H₅F₃O
MW: 162.11
MDL Number: MFCD00042397
CAS Number: 203245-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR