3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide 98%

Supplier: Ambeed
Warning

Synonyms: Antioxidant 1024, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)propanehydrazide, 1,2-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazine

A594133-100G A594133-25G
77005-340EA 20.16 USD
77005-340 77005-342
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide 98%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide

Caution: for Research Use Only

Formula: C₃₄H₅₂N₂O₄
MW: 552.80 g/mol
MDL Number: MFCD00072662
CAS Number: 32687-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR